Personna Plus® Microtome Blades
Supplier: Avantor
Certificates
About this item
Engineered to perform.
- Available in High Profile or Low Profile formats
- Round corners
- Safety dispenser, used blade vault and round corner blades
- 3 facet edge
- Made in U.S.A
There is no room for error when sampling and sectioning. These stainless steel microtome blades are precision ground with evenly matched and aligned blade edges and optimal cutting angles. The propietary MicroCoat® surface produces smoother cuts with less compression and greater section clarity.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together