Order Entry
ContactUsLinkComponent
Human NAP-2 (CXCL7) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Human NAP-2 (CXCL7) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Catalog #: 10777-680
Supplier:  Peprotech
CAS Number:  
Human NAP-2 (CXCL7) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Catalog #: 10777-680
Supplier:  Peprotech
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Reactivity:
    Human
  • Description:
    Human NAP-2 (CXCL7) ELISA Kit
  • Size:
    Kit components sufficient to assay 10 plates
  • Cat. no.:
    10777-680
  • Supplier No.:
    900-K40

Specifications

About this item

PeproTech's Human NAP-2 (CXCL7) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human NAP-2 (CXCL7) in a sandwich ELISA format.

Easy-to-use
Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)

Human NAP-2 (CXCL7) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human NAP-2 (CXCL7) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human NAP-2 (CXCL7) in approximately 1000 ELISA plate wells.

Customers Who Bought This Also Bought