Spouting Cylinder
Catalog #: 470303-934
Supplier: United Scientific Supplies
Specifications
- Cat. no.:470303-934
- Dimensions:Height: 195/16"
Specifications
About this item
Demonstrate Torricelli’s theorem to show the relationship between water pressure and depth.
- Clear design
- Three nozzles at different heights
This clear acrylic cylinder includes three vertically aligned spouting nozzles positioned 43/4" apart.
Each spouting nozzle also includes a stopper so pressure at each height can be shown individually.
A filler hose barb at the top can be used to quickly refill the cylinder or maintain a constant water depth.
Customers Who Bought This Also Bought