Sterilization Pouch Rack, Sklar®
Supplier: Sklar
Certificates
About this item
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The Sterilization rack has a coil design to separate and hold sterilization pouches used with steam sterilization, EO Gas, and Gas Plasma.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.