Specifications
- Length:5.5"
- Description:Fox Dermatology Curette
- Material:Stainless Steel
- Tip Configuration:Round
- Cat. no.:76191-914
- Handle:Flat
Specifications
About this item
This is used for scraping skin lesions and growths such as warts or melanomas.
- Industry standard
- The handle is lightweight
It is a single-end instrument available with a loop tip. The loop is sharp on the inside and blunt on the outside so the lesion can be removed but surrounding skin is not damaged.