About this item
Secure specimens during processing with the Unisette™ Biopsy Processing/Embedding Cassette.
- In Quickload™ stacks
- 35° anterior writing area
- One-piece construction with hinge-lock design prevents early separation of base and lid
- Snap latch and hinge-lock design; cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.