Fmoc-1,6-diaminohexane hydrochloride ≥95%
Supplier: CHI Scientific
Certificates
About this item
Synonyms: N-1-Fmoc-1
Formula:C₂₁H₂₇ClN₂O₂
MW:374.909 g/mol
MDL Number:MFCD00830743
CAS Number:166410-37-3
:Storage Temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 374.909 g/mol
Storage Temperature:Freezer
Specification Test Results
- Protease-free:Yes
- Animal-free:Yes
- Endotoxin Level:low (0.1 ng/μg)
Specification Test Results
Customers Who Bought This Also Bought