To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme type:Recombinant
- Packaging:Plastic Ampoule
- Source:Aspergillus niger
- Size:10 KU
- Storage conditions:2...8 °C
- Enzyme name:Glucose Oxidase
- Cat. no.:CA80602-310
- Supplier No.:345386-10KU
Specifications
About this item
Recombinant, Aspergillus niger glucose oxidase catalyzes the oxidation of β-D-glucose to D-glucono-ω-lactone and hydrogen peroxide and also catalyzes the conversion of D-glucono-ω-lactone to gluconic acid. Used for enzymatic assay of glucose concentration.
- Activity: ≥250 units/mg solid
- Form: Yellow-brown lyophilized solid
One unit is defined as the amount of enzyme that will oxidize 1.0 mmol of β-D-glucose per minute at 25 °C, pH 7.0.