About this item
Made of acetal and resistant to the chemical action of most histological solvents and suitable for automated printers.
- In Quickload™ stacks, for Primera printers
- 45° anterior writing area
- 1676 square openings (0.36 mm); no biopsy pads needed
- One large 27 mm compartment
- Recessed cover
Secure specimens during processing with Micromesh™ Biopsy Processing/Embedding Cassettes. No need for biopsy pads thanks to the 1676 square 0.36mm openings of the mesh.