Specifications
- Length:7"
- Description:Wolff Cast Breaker
- Material:Stainless Steel
- Cat. no.:76191-614
- Jaw Type:Curved
Specifications
About this item
Sklar's® Wolff Cast Breaker is used for breaking apart plaster casts.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
It is a single spring, plier instrument with jaws the same length as the shanks. The jaws are slightly curved and have deep horizontal serrations half the length the jaws. The tips are blunt.