Specifications
- Length:6"
- Description:Ring Cutter, Sharp Tip
- Material:Stainless Steel
- Cat. no.:76191-454
Specifications
About this item
Sklar's® Ring Cutter is a speciality instrument specifically used for cutting and removing rings from a traumatized or otherwise inflamed finger.
It features a thumb screw that turns a sharp, toothed, rotary blade that cuts the ring. The single, grooved handle features a thumb lever which operates a lower jaw that slides under the ring and protects the finger from the cutting.
This ring cutter is best used on hard metals such as titanium and steel.