To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Immunoglobulin G2b
- Clonality:Monoclonal
- Clone:LN-3
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Paraffin sections
- Antigen symbol:IgG2b
- ImmunoFluorescence:Yes
- Size:2 ml
- Form:Liquid
- Storage buffer:pH: 7
- Storage temperature:2-8°C. Stability of liquid at 2-8°C is at least one year. If stored frozen (15°C or below) for long term, avoid repeated
thawing and freezing - Shipping temperature:4C
- Cat. no.:IC08693031
Specifications
About this item
LN-3 is a murine monoclonal antibody (IgG2b) reactive with a nonpolymorphic HLA-DR (Ia-like) antigen. The
antigens show the typical 29-33 K dalton molecular weight of Ia. The HLA-DR antigen is present on germinal center and mantle
zone B cells, monocytes and macrophages and interdigitating histiocytes. T cells are generally negative although activated T
cells may express HLA-DR
Type: Primary
Antigen: IgG2b
Clonality: Monoclonal
Clone: LN-3
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Paraffin Sections