Anti-Sheep Serum Goat Polyclonal Antibody
Catalog #: IC0856006
Supplier: MP Biomedicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Goat
- Reactivity:Sheep
- ELISA:Yes
- Size:2 ml
- Format:Lyophilized
- Form:Lyophilized powder
- Storage buffer:pH: 7.3
- Storage temperature:Store product at 2-8°C. Store reconstituted material no longer than 4 weeks at 2-8°C. For long-term storage, aliquot and freeze
reconstituted product at –20 °C or lower. - Concentration:Total Protein: 62.6 mg/mL
- Shipping temperature:4C
- Cat. no.:IC0856006
Specifications
About this item
Product is the lyophilized powder of goat antiserum to sheep serum and buffer salts.
Type: Primary
Antigen: Sheep Serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Sheep
Customers Who Bought This Also Bought