To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Goat
- Reactivity:Human
- Size:2 ml
- Format:Lyophilized
- Form:Lyophilized powder
- Storage buffer:pH: 7.3
- Storage temperature:Store product at 2-8 °C. Store reconstituted material no longer than 4 weeks at 2-8 °C. For long-term storage, aliquot and freeze reconstituted product at -20 °C or lower.
- Shipping temperature:4C
- Immunogen:Human Serum
- Cat. no.:IC0855996
Specifications
About this item
Product is the lyophilized powder of goat antiserum to human serum and buffer salts.
The antiserum is prepared by immunizing goats with human serum; antiserum is delipidated and made specific by
immunoadsorption with soluble antigens or solid-phase immunoadsorption where appropriate. The resulting antiserum is
dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer,
filtered through a 0.22 µm filter, vialed and lyophilized
Type: Primary
Antigen: Human Serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human serum