UVP Transilluminator, Visi-Blue™, Analytik Jena
Supplier: Analytik Jena CA
Certificates
About this item
The UVP Visi-Blue Transilluminator features 8 watt or 25 watt models with 21×26 cm and 20×40 cm filter sizes respectively.
Benefits of these transilluminators include high intensity and uniformity, instant on, no lamp flicker, reduced electrical consumption, long-life filter, and a Visi-Blue cover.
Certifications: CSA certified.
Ordering information: Two-year warranty.
Customers Who Bought This Also Bought