96-Well Reaction Plates, Chemglass
Supplier: Chemglass
Certificates
About this item
The reaction plate is made of 33 expansion borosilicate glass.
- Wells are coded with letters A to H vertically and numbers 1 to 12 horizontally
- Autoclavable
- Designed for use with chemistry applications and resistant to temperatures up to 530 °C
Overall dimensions: 85×127 mm
The plates are automated from a homogeneous glass sheet also feature uniform geometry and shape to provide consistent temperatures from well to well within 0.2 °C typical.
The uniformity of the thickness of the plate bottoms is approximately ±0.1 mm and the bottom is frosted, these plates are not recommended for use with optical detection screening experiments.
Customers Who Bought This Also Bought