Condensate Recirculators
Supplier: CARON PRODUCTS & SERVICES, INC. MX
Certificates
About this item
These recirculators acts as a water delivery system for use with any Caron humidity controlled chamber.
- Environmentally friendly
- Dimensions: 22.8W×43.1L×17.7cm H (9×17×7")
- Durable powder coated metal as external material
- Water reservoir 7 gal.
- Water purity 100 to 125 KΩ.cm
These are designed to utilize tap water, eliminating the need to purchase more expensive water purification systems or maintain a supply of distilled or deionized water.
The single switch operation and automatic air purge minimize and simplify user interaction. The front/rear handles and casters provide easy mobility for installation and service.
The optional UV germicidal lamp deactivates microorganisms including bacteria, molds, protozoa, viruses and yeasts for contamination-free water.
Customers Who Bought This Also Bought