VWR® Economy Digital Thermometer
Catalog #: 89140-510
Supplier: VWR International
Specifications
- Resolution:0.1
- Description:VWR® Digital Thermometer with Probe
- Probe:Sensor with 39" immmersible cable
- Range:–10...50 °C (14...122 °F), ambient; –50...70 °C (–58...158 °F), probe
Specifications
About this item
Economy digital thermometer with probe and used in ambient temperature.
- High or low alarm throughput
- With minimum and maximum memory
- Available in °C and °F
Customers Who Bought This Also Bought