SCIENCE-MARKER™ Cryogenic Markers, GA International
Supplier: GA INTERNATIONAL INC
Certificates
About this item
Cryogenic permanent marker with fine tip.
- Alcohol and water resistant
- Compatible with Lab-TAG™ labels
Shipping weight 113 g
Delivery: Supplied six markers per pack.
Caution: Not suitable for exposure to Acid/Ethanol mixes.
Customers Who Bought This Also Bought