2,3,5-Triphenyltetrazolium chloride (TTC) ≥98% (by perchloric acid titration) for microbiology
Catalog #: CA1.08380.0100
Supplier: MilliporeSigma
CAS Number: 298-96-4
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:100 g
- Cat. no.:CA1.08380.0100
- Supplier No.:1.08380.0100
Specifications
Specification Test Results
- Assay (perchloric acid titration, calculated on dried substance):≥98%
- Identity (UV/VIS-Spectrum):Passes Test
- Absorption maximum λmax. (water):245 - 248 nm
- Spec. Absorptivity A 1%/1cm (λmax; 0,01 g/L; water; calculated on dried substance):≥770
- TLC-Test:Passes Test
- Loss on drying (70 °C; ≤1 hPa):≤0.5%
- Suitability for testing the germinability of seeds:Passes Test
- Suitability for microbiology:Passes Test
- 2,3,5-Triphenyltetrazolium chloride (TTC) ≥98% (by perchloric acid titration) for microbiology:
Specification Test Results
About this item
Synonyms: Tetrazolium salt,Tetrazolium Red,2,3,5-Triphenyl-2H-tetrazol-3-ium chloride,2,3,5-Triphenyl-2H-tetrazolium chloride
Melting Pt:243 °C (dec.)
ADR:4.1,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 243 °C (dec.)
:Flash Pt:
:Flash Pt:
:M.W. 334.81 g/mol
:2,3,5-Triphenyltetrazolium chloride (TTC)
:243 °C
Formula:C₁₉H₁₅ClN₄
MW:334.81 g/mol
MDL Number:MFCD00011963
CAS Number:298-96-4
UN:1325
:4.1
:II
:GHS02
:GHS07
:12,09874
Customers Who Bought This Also Bought