About this item
FOXP3 is a 50-55 kD transcription factor, also known as Forkhead box protein P3, Scurfin, JM2, or IPEX. It is proposed to be a master regulatory gene and more specific marker of T
regulatory cells than most cell surface markers (such as CD4 and CD25).
Transduced expression of FOXP3 in CD4 /CD25 cells has been shown to induce GITR, CD103, and CTLA4 and impart a T regulatory cell phenotype. FOXP3 is mutated in X-linked
autoimmunity-allergic dysregulation syndrome (XLAAD or IPEX) in humans and in "scurfy" mice.
Overexpression of FOXP3 has been shown to lead to a hypoactive immune state suggesting that this transcriptional factor is a central regulator of T cell activity. In human,
unlike in mouse, two isoforms of FOXP3 have been reported: one (FOXP3) corresponding to the canonical full-length sequence; the other (FOXP3 δ2) lacking exon 2. The 206D antibody recognizes human FOXP3 epitope in the region of amino acids 105-235.
Type: Primary
Antigen: FOXP3
Clonality:
Clone: 206D
Conjugation: Brilliant Violet 421™
Epitope: 105-235
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Cynomolgus, Rhesus, Baboon, Pigtailed Macaque
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA75780-626
- CA75780-380
- Antigen name:
- Forkhead Box P3
- Forkhead Box P3
- Antibody type:
- Primary
- Primary
- Clone:
- 206D
- 206D
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 50943, 50943, 20371, 317382, 916
- 50943, 50943, 20371, 317382, 916
- Isotype:
- IgG1 kappa
- IgG1 kappa