TLC Plate Rack, Kimble Chase, DWK Life Sciences
Catalog #: KT416175-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:TLC aluminum rack
- Cat. no.:KT416175-0000
- No. of Places:6
- Supplier No.:416175-0000
Specifications
About this item
TLC plate racks for use with the TLC rectangular developing tanks.
- Holds 20×20 cm plates
Customers Who Bought This Also Bought