Basic Industrial Comfort Waist Pants, Dickies®
Supplier: Dickies
Certificates
About this item
Dickies® style LP817.
- Stay Dark® technology, wrinkle resistant, stain release
- Enhanced durability, wider and stronger belt loops
- Hidden expandable comfort waistband, button closure
- Reinforced, deep front pockets
- Left back pocket has button closure
- 7.75 oz. vat dyed twill, 65% polyester/35% cotton
Sizing Chart: UU size designations represent a 37" inseam, unhemmed. UL size designations represent a 39" inseam, unhemmed.
Specifications
- Catalog No:
- 76154-834
- 76154-836
- 76154-830
- 76154-918
- 76154-832
- 76154-914
- 76154-826
- 76154-828
- 76154-916
- 76154-910
- 76154-822
- 76154-912
- 76154-824
- 76154-906
- 76154-908
- 76154-820
- 76154-902
- 76154-904
- 76154-900
- 76154-838
- 76154-658
- 76154-896
- 76154-654
- 76154-898
- 76154-656
- 76158-274
- 76158-272
- 76154-892
- 76154-650
- 76154-652
- 76154-894
- 76158-270
- 76154-888
- 76154-646
- 76158-268
- 76154-890
- 76154-648
- 76154-642
- 76154-884
- 76154-818
- 76154-644
- 76154-886
- 76154-814
- 76154-880
- 76154-882
- 76154-816
- 76154-640
- 76154-810
- 76154-812
- 76154-806
- 76154-808
- 76154-878
- 76154-638
- 76154-874
- 76154-876
- 76154-870
- 76154-872
- 76154-866
- 76154-868
- 76154-796
- 76154-862
- 76154-798
- 76154-864
- 76154-792
- 76154-794
- 76154-860
- 76154-788
- 76154-790
- 76154-802
- 76154-804
- 76154-800
- 76154-856
- 76154-858
- 76154-852
- 76154-854
- 76154-848
- 76154-850
- 76154-844
- 76154-846
- 76154-840
- 76154-842
- 76154-924
- 76154-920
- 76154-922
- Model:
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- LP817
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- Weight:
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.
- 7.75 oz.