SP Bel-Art Benchtop Biohazard Disposal Can with Motion Sensor Lid, Bel-Art Products, a part of SP
Catalog #: 76226-064
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:1.5 L
- Cat. no.:76226-064
- Diameter ×H:12.4 × 17 cm (44/5 × 67/10")
Specifications
About this item
Compact benchtop disposal can contains odors and is perfect for small items such as pipette tips and microcentrifuge tubes.
- Automatic opening lid provides hands-free use; closes automatically and securely for a more sanitary environment
- Holds a 25×25 cm (10×10") autoclavable biohazard disposal bag (H13182-0000)
- Includes a polypropylene liner with handle for easy emptying of contents
- Aesthetic and durable; stainless steel body and chrome lid are resistant to bleach and easy to clean
Certifications: CE and RoHS.
Ordering information: Requires (3) AAA batteries (not included).
Customers Who Bought This Also Bought