Kilo Scale Bubbler, Ace Glass
Catalog #: 76214-458
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Width:175 mm
- Description:Kilo scale bubbler with 0 - 10 mm valve
- Height:280 mm
- Cat. no.:76214-458
- Depth:210 mm
Specifications
About this item
Kilo scale bubbler for inerting or purging large reaction vessels.
- High-quality borosilicate glass
- Ace-Safe™ hose connections eliminate the safety hazard of broken glass hose connections
Bubbler features a ground glass check valve which prevents oil or air from being pulled into the system.
Ace-Safe™ hose connections for 3/8" ID tubing.
Customers Who Bought This Also Bought