Distilling Head, Ace Glass
Catalog #: 76214-410
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Description:Distillation Head
- Cat. no.:76214-410
- Thermometer ST Joint Size:14/20
- Effective Length:250 mm (condenser)
Specifications
About this item
Distilling head sits on top of your vapor path column. Has condenser side arm with drip tip.
- High-quality borosilicate glass
- 24/40 standard taper bottom joint and side arm joint
- #11 Ace-Thred inlet/outlet for use with Ace-Safe hose connections
Can be joined with Ace short stem vacuum take-off adapters.
Customers Who Bought This Also Bought