Order Entry
ContactUsLinkComponent
SP Wilmad-LabGlass Distilling Adapter with 75° Angle Side Arm, SP Industries
SP Wilmad-LabGlass Distilling Adapter with 75° Angle Side Arm, SP Industries
Catalog #: 14203-512
Supplier:  SP Industries
Adapter, Distillation, 75[degree]
SP Wilmad-LabGlass Distilling Adapter with 75° Angle Side Arm, SP Industries
Catalog #: 14203-512
Supplier:  SP Industries

Specifications

  • ST Joint:
    24/40
  • Description:
    75 mm Immersion Length
  • Cat. no.:
    14203-512
  • Thermometer ST Joint Size:
    10/30
  • Supplier No.:
    LG-1530-100

Specifications

About this item

This borosilicate glass three-way adapter has a Standard Taper 10/30 outer joint on top to accommodate a thermometer, bottom inner Standard Taper 24/40 joint, and 75° angle inner side joint of same size.

Customers Who Bought This Also Bought