Sealing Films, Sealplate, Chemglass
Supplier: Chemglass
Certificates
About this item
Polyester sealing films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal. Secure sealing of all wells eliminates edge effects in sensitive ELISA assays.
- Functional temperature range from –40 to 120 °C
- Available sterile or non-sterile
- Sealing films are non-pierceable
SealPlate® sealing films can also be used with tissue culture plates for short-term storage, incubation, and containment of biohazards.
Each SealPlate® sealing film measures 79.4×146.1 mm.
Packaging: Packed in boxes of 100.
Customers Who Bought This Also Bought