Top Dog 8 Mil Die Cut Apron, 45" Length, Remco Products
Supplier: Remco Products
Certificates
About this item
Reusable and washable, the one-piece construction increases the apron's hygiene and durability.
- Resistant to chemical and acid sanitation products
- Non-porous surface will not host bacteria or mold
- Washer and dryer safe
- Will not crack or stiffen in low temperatures or with repeated cleaning
- Impervious to animal fats and degreasers
- Completely latex-free for sensitive users
- Easily repairable
8 mil aprons from Top Dog come in five colors to support color-coded HACCP plans and zoning efforts. The 8 Mil Die Cut Apron is made of pure polyurethane and is reversible. It fits comfortably over clothes. The apron is machine washable, dryer-safe to 180 °F, and chemical and abrasive resistant. The 45" length makes the apron cover most of an employee's torso and legs without becoming cumbersome.
Certifications: Made of FDA Compliant material.
Ordering information: Aprons can only be purchased by the case. Case quantity is 24.
Delivery: Dropshipping available.
Specifications
- Catalog No:
- 76196-580
- 76196-574
- 76196-572
- 76196-578
- 76196-576
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Fabric:
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Thickness:
- 8 mil
- 8 mil
- 8 mil
- 8 mil
- 8 mil
Specifications
Customers Who Bought This Also Bought