Self-Evacuating Cash Still with Teflon® Stopcock, General Glass Blowing
Catalog #: 89107-064
Supplier: General Glass Blowing
Specifications
- Cat. no.:89107-064
- Supplier No.:GW105-S
Specifications
About this item
The Self Evacuating Cash Still complies with the AOAC method to determine Acidity (Total Volatile) of wines. The steam chamber is heated by an 800 watt immersion coil that heats the distilled water to a boiling point in about 3 to 4 minutes.
- Made of borosilicate glass
Two way Teflon stopcock fitted with funnel, permits addition of sample into inner chamber or serves as an escape vent for CO2. The Graham condenser has threaded fittings on the water feed and drain that makes the assembly and disassembly of the unit easier.
Ordering information: Consists of: Outer chamber, inner chamber, Graham condenser, electric coil heater and 3-way Teflon® stopcock.
Customers Who Bought This Also Bought