Dextran sulfate sodium salt solution 50%, OmniPur®, sterile, Calbiochem®
Catalog #: CA3730
Supplier: MilliporeSigma
CAS Number: 9011-18-1
Specifications
- Packaging:Plastic bottle
- Size:100 ml
- Supplier No.:3730
Specifications
Specification Test Results
- DNase:None detected
- Protease:None detected
- RNase:None detected
- Calbiochem®, Dextran sulfate sodium salt solution 50%, OmniPur®, sterile:
Specification Test Results
About this item
Synonyms: Dextran sulfate sodium from Dextran
:Storage Temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:Dextran sulfate sodium salt
Formula:(C₆H₇Na₃O₁₄S₃)n
Storage Temperature:Ambient
MDL Number:MFCD00081551
CAS Number:9011-18-1
Customers Who Bought This Also Bought