ATP and Cell Energy Model Set
Catalog #: 470232-066
Supplier: Mega Molecules
Specifications
- Description:ATP and Cell Energy
- Cat. no.:470232-066
Specifications
About this item
Demonstrate ADP and ATP.
- Great model for high school and college students
ATP transports chemical energy within cells for metabolism. One molecule of ATP contains three phosphate groups, and it is produced from inorganic phosphate and adenosine diphosphate (ADP) or adenosine monophosphate (AMP). Red Links are used to simulate the high-energy phosphate bonds. This model may be used to illustrate that metabolic processes use ATP as an energy source and convert it back into organic phosphate and ADP.
Customers Who Bought This Also Bought