Balancing Act!
Catalog #: 470232-060
Supplier: Mega Molecules
Specifications
- Description:Balancing Act!
- Cat. no.:470232-060
Specifications
About this item
What is the resulting balanced equation?
- Perfect for middle and high school students
Balancing act makes learning to balance chemical equations fun and easy. Students practice balancing chemical equations using hands-on molecular models to simulate combustion reactions. Students identify the reactants (starting materials) and products (resulting substance). This set makes it easy to count the number of atoms of each element and see the mathematical relationship between the quantity of reactants and products.
Customers Who Bought This Also Bought