ContactUsLinkComponent
Total Immersion Alcohol Thermometers
Total Immersion Alcohol Thermometers
Catalog #: 470104-556
THERM TOTAL IMMER RED ALC -20-110C 30CM
Total Immersion Alcohol Thermometers
Catalog #: 470104-556
Specifications
  • Length:
    30 cm (12")
  • Description:
    Red Alcohol Filled
  • Divisions:
    1 °C
  • Range:
    −20…+110 °C
  • Immersion:
    Total
  • Supplier No.:
    M80232A
  • Background Color:
    White
Specifications
About this item
Accurate, economical, and easy to use.
  • Red alcohol thermometer
  • Polypropylene sleeve packaging included
Customers Who Bought This Also Bought