Specifications
- Length:4.5"
- Material:Stainless Steel
- Tip Configuration:2 x 3 Prongs, Blunt
- Cat. no.:76192-462
Specifications
About this item
The Econo™ Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Econo™ Weitlaner Retractor has curved shanks that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed. The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries. It is also useful with small, deep incisions and soft tissue dissection at a superficial level. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.