About this item
The Sklar® Castroviejo needle holders are used for micro surgery, for suturing and stapling.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Castroviejo needle holders hold very fine suture needles in opthalmic procedures. Castroviejo's can be opened from any rotational position, and are available straight or curved. The diamond jaw serration prevents rotation of the needle. Sklar® permanently bonds tungsten carbide inserts to the instrument, ensuring shaprness for a longer period of time, and maximizing control and precision. The tungsten carbide line of instruments is identified by gold handles.