Specifications
- Length:7"
- Material:Stainless Steel
- Tip Configuration:2 Prongs, Sharp
- Working Surface Style:Pointed
- Curvature:Straight
- Cat. no.:76192-824
Specifications
About this item
The Sklar® Guthrie Hook is a double-prong tissue hook used for handling and retracting tissue.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Sklar® Guthrie Hook is often used in dermatologic and plastic surgery procedures, but the tiny hooks also make this retractor useful in ophthalmologic and intranasal procedures. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities. They eliminate the need for cleaning and reprocessing, and reduce the likelihood of cross-contamination.