Specifications
- Length:7.25"
- Material:Stainless Steel
- Curvature:Slightly Curved
- Cat. no.:76192-636
Specifications
About this item
The Econo™ Freer Elevator is an orthopedic instrument that can be used as a multipurpose tool in many procedures.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Econo™ Freer Elevator is primarily used for blunt debulking and lifting periosteum from bones in confined areas. This double-ended instrument has a rounded handle with curved, tear-drop tips, one sharp and one blunt. Econo™ line instruments are floorgrade, disposable instruments sold in bulk quantities.