Bizweld Flame Resistant Cargo Pants, Portwest
Supplier: Portwest
Certificates
About this item
Dual hazard protection, guaranteed flame resistance for life of garment.
- ARC2, 11.2 cal./cm²
- 2 side pockets, 1 leg pocket, 1 concealed cell phone pocket, 2 rear pockets, 1 rule pocket
- Elasticated waistband for extra comfort
- Blue and Gray color options
- 31" inseam
Certifications: ASTM F1959/F1959M-12, ATPV 11.2 cal./cm² (HAF 80.4%), ASTM F1506-10A, NFPA 70E, NFPA 2112.
Specifications
- Catalog No:
- 76103-822
- 76103-832
- 76103-590
- 76103-592
- 76103-834
- 76103-594
- 76103-948
- 76103-824
- 76103-826
- 76103-828
- 76103-588
- 76103-830
- 76103-950
- 76103-952
- 76103-954
- 76103-956
- Garment application:
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Fabric:
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
- Bizweld (100% FR treated cotton)
Specifications
Customers Who Bought This Also Bought