Bizflame 88/12 Flame Resistant Shirts, FR89, Portwest
Supplier: Portwest
Certificates
About this item
This stylish shirt is lightweight and comfortable. Maximum protection is guaranteed with permanent flame resistance and ARC2 protection against electric arc.
- Dual hazard protection
- Guaranteed flame resistant for life of garment
- 2 front pockets, pen division on left pocket
- Button cuffs for a secure and comfortable fit
- Shirt tail hem stays tucked in
- 40+ UPF rated fabric to block 98% of UV rays
Made from Bizflame 88/12 fabric: 88% Cotton, 12% Nylon.
Certifications: ASTM F1959/F1959M-12, ATPV 8.2 cal./cm² (HAF 69.1%), ASTM F1506-10A, NFPA 70E, NFPA 2112.
Specifications
- Catalog No:
- 76104-298
- 76104-300
- 76104-112
- 76104-310
- 76104-114
- 76104-312
- 76104-292
- 76104-294
- 76104-302
- 76104-304
- 76104-306
- 76104-110
- 76104-308
- 76104-296
- Garment application:
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Industrial, Fire protection, Electronics/semi-conductor
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
Specifications
Customers Who Bought This Also Bought