About this item
Econo™ Fox Dermal Curettes are used for scraping skin lesions and growths such as warts or melanomas.
- Variety of tip sizes are also available to accommodate varying lesion sizes
- Industry standard
This is a single-ended instrument available with an oval or round loop tip. The loop is sharp on the inside and blunt on the outside so the lesion can be removed but surrounding skin is not damaged.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76192-380
- 76192-378
- Disposable / reusable:
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free