Sealex Flame Resistant Hi-Vis Weatherproof Jackets and Pants, Portwest
Supplier: Portwest
Certificates
About this item
Garments offer protection against adverse weather conditions and flame hazards.
- Flame resistant, treated, waterproof 7.5 oz. fabric
- 2" flame resistant reflective tape
- Zipper storm flap front closure
- Pack away adjustable hood
- Easy access cargo pockets
- Snap-on cuffs for snug fit
- Welded waterproof seams
Certifications: ANSI/ISEA 107-2015 Class 3.
Specifications
- Catalog No:
- 76104-090
- 76104-092
- 76104-094
- 76104-096
- 76104-082
- 76104-084
- 76104-086
- 76104-088
- 76104-098
- 76104-100
- Garment application:
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Fire protection, Outdoor
- Risk area:
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Weight:
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
- 7.5 oz.
Specifications
Customers Who Bought This Also Bought