DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Open Wrists and Ankles, Serged Seams
Supplier: DuPont
Certificates
About this item
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
- Laydown collar
- Open wrists and ankles
- Serged seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls feature a laydown collar, open wrists, and open ankles.
Certifications: Thickness meets ASTM D1777.
Specifications
- Catalog No:
- CA10010-190
- CA10010-192
- CA10010-186
- CA10010-188
- CA10010-182
- CA10010-184
- CA13452-630
- 10016-366
- 10016-368
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory