Protected Ambient Category B Shipping Systems, Therapak®
Supplier: Therapak, LLC
Certificates
About this item
Protected ambient category B shipping Systems are ideal for shipping diagnostic specimens.
- Compact, easy to use
- Category B certified
- Interchangeable Category B and EHS labeling
- Includes Aqui-Pak™ absorbent pouch
Whether using active or passive forms of temperature spike buffering, these shippers offer a higher level of protection during shipment than shipping specimens alone. All three systems come with both a Category B and EHS label for easy interchangeability. The systems come with 95 kPa secondary packaging.
Certifications: Meets IATA Packing Instructions 650 requirements to transport Category B Specimens.
Customers Who Bought This Also Bought