Dupont™ ProShield® 60 Coveralls with Standard Hood and Attached Skid Resistant Boots
Supplier: DuPont
Certificates
About this item
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- Standard fit hood
- Elastic wrists
- Attached skid-resistant boots
- Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Attached hood with elastic around face opening is standard fit. Front zipper closure allows for easy donning and doffing. Elastic opening at wrists allows for a tighter fit. Attached ProShield® 3 boots have a special coating to provide added skid-resistance.
Certifications: Thickness meets ASTM D751.