ContactUsLinkComponent
Carson eFLEX Digital Microscope
Carson eFLEX Digital Microscope
Catalog #: 470235-858
Supplier:  CARSON OPTICAL
Eflex Digital Microscope
Carson eFLEX Digital Microscope
Catalog #: 470235-858
Supplier:  CARSON OPTICAL
Specifications
  • Description:
    eFlex
  • Model:
    MM-840
  • Computer requirements:
    Compatible with Windows 10/8/8.1/7/Vista/XP SP2+ and all Mac OS. A USB 2.0/3.0
  • Cat. no.:
    470235-858
Specifications
About this item
The eFlex from Carson is a powerful digital microscope that displays the magnified image right on your computer screen.
  • 1600 x 1200 max picture resolution
  • 640 x 480 max video resolution
The impressive 75x/300x effective magnification (on 21 inch monitor) allows you to see details of ordinary objects that you never knew existed.
Capture an image to keep with the click of a button. Captures close-focus video. The included flexible neck stand with suction cup adapter attached to any smooth surface, or included graduated base with ease.
Caution: Does not work with tablets.
Customers Who Bought This Also Bought