ContactUsLinkComponent
Sharps Containers, Stackable, Medegen
Sharps Containers, Stackable, Medegen
Catalog #: 89166-424
Medical Action* Stackable Sharps Containers 3gal
Sharps Containers, Stackable, Medegen
Catalog #: 89166-424
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Capacity:
    11.35 L (3 gal.)
  • Color:
    Red
  • Description:
    Sharps Container with Slide Lid
  • Cat. no.:
    89166-424
  • Supplier No.:
    8710
Specifications
About this item
Large multi-purpose sharps containers are suitable for OR and dialysis units, and contain locks for final disposal.
Containers feature a tortuous path lid with large and small openings for infectious waste of various types.
Customers Who Bought This Also Bought