About this item
UACA (Uveal Autoantigen with Coiled-coil domains and Ankyrin repeats) is a 1,416 amino acid nuclear membrane protein. It was originally identified as an autoantigen in patients with panuveitis, a characteristic of Vogt-Koyanagi-Harada disease, and in patients with Graves' disease. UACA was also later identified as Nucling, an mRNA differentially expressed in F9 embryonal carcinoma cells during cardiac muscle differentiation. UACA appears to function as a pro-apoptotic protein that recruits the apaf-1-pro-caspase-9 complex for the induction of apoptosis to mediate the cell-death pathway.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Type: Primary
Antigen: UACA
Clonality: Monoclonal
Clone: Clone UACA/1222
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75978-854
- 75978-852
- Antigen name:
- UACA / Nucling
- UACA / Nucling
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- UACA/1222
- UACA/1222
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 108049
- Unigene ID: 108049
- Isotype:
- IgG1 kappa
- IgG1 kappa