To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neurofilament
- Clonality:Monoclonal
- Clone:RT-97 NR-4
- Gene ID:Unigene ID: 198760
- Host:Mouse
- Isotype:IgG
- Reactivity:Pig,Chicken,Human,Rat,Mouse
- Antigen symbol:NEFH & NEFL
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:50 μL
- Storage buffer:PBS, no BSA, no azide
- Concentration:1 mg/mL
- Cat. no.:75965-848
- Supplier No.:BNUM1201-50
Specifications
About this item
This MAb reacts with a 200 kDa and 68 kDa protein, identified as heavy and light sub-units of neurofilaments (NF-H & NF-L). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68 kDa (NF-L), 160 kDa (NF-M) and 200 kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament.
Type: Primary
Antigen: NEFH & NEFL
Clonality: Monoclonal
Clone: Clone RT-97 NR-4
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Chicken, Human, Mouse, Pig, Rat