About this item
HLA-A, with HLA-B and HLA-C, belongs to major histocompatibility complex (MHC) class I antigens and expresses constitutively on all nucleated cells. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation. This MAb is useful for HLA molecular typing of peripheral blood leukocytes as well as a large number of leukemic cell lines. It reacts with an intralocus determinant present on a limited number of HLA-A locus-encoded gene products (HLA-A2, -A3, -A28, -A29, -A30, -A31 and -Aw33). Its epitope maps between aa65-to-aa80 of thealpha1 domain of the HLA-A. This MAb recognizes an intralocus determinant present on a limited number of HLA-A locus-encoded gene products (HLA-A2, -A3, A28, -A29, -A30, -A31 and -Aw33). Furthermore, by testing its reactivity with HLA-A2 natural variants and mutants, the importance of amino acid residues 79 and/or 80 of thealpha1 domain was demonstrated in the formation of an intralocus HLA-A determinant.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.
Type: Primary
Antigen: HLA-A
Clonality: Monoclonal
Clone: Clone 108-2C5
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75962-716
- 75962-718
- Antigen name:
- MHC I
- MHC I
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 108-2C5
- 108-2C5
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 181244; 654404; 77961
- Unigene ID: 181244; 654404; 77961
- Isotype:
- IgG1 kappa
- IgG1 kappa